Assay Details
Target Gene Details
Entrez Gene ID: | 2962 |
Gene Name: | general transcription factor IIF subunit 1 |
Gene Aliases: |
BTF4, RAP74, TF2F1, TFIIF |
Location: |
Chr.19:6379569-6393280 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTF2F1 | NM_002096.2 | 13 | 1809 | NP_002087.2 |
| AK293124.1 | 13 | 1409 | ||
| AK302178.1 | 10 | 1272 | ||
| AK315240.1 | ||||
| BC000120.1 | 13 | 1671 | AAH00120.1 | |
| BC013007.2 | 13 | 1671 | AAH13007.1 | |
| BT007097.1 | AAP35761.1 | |||
| CR456798.1 | CAG33079.1 | |||
| X64002.1 | 13 | 1587 | CAA45404.1 | |
| X64037.1 | 13 | 1702 | CAA45408.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv513757 | Chr.19:6375553 - 6388070 on Build GRCh38 | Insertion |
|
| esv3643556 | Chr.19:6377364 - 6387673 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map