Assay Details
Target Gene Details
Entrez Gene ID: | 50619 |
Gene Name: | DEF6, guanine nucleotide exchange factor |
Gene Aliases: |
IBP, SLAT, SWAP70L |
Location: |
Chr.6:35297818-35321771 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DEF6 | NM_022047.3 | 8 | 1378 | NP_071330.3 |
| AJ276095.1 | 8 | 1339 | CAC08450.1 | |
| AK093613.1 | 7 | 1090 | ||
| AK314756.1 | 8 | 1353 | ||
| AY241694.1 | 8 | 1378 | AAO91767.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759419 | Chr.6:35200339 - 35323225 on Build GRCh38 | Loss |
|
| nsv830641 | Chr.6:35314757 - 35472826 on Build GRCh38 | Loss |
|
| nsv602907 | Chr.6:35317943 - 35344490 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map