Assay Details
Target Gene Details
Entrez Gene ID: | 57221 |
Gene Name: | ARFGEF family member 3 |
Gene Aliases: |
A7322, BIG3, C6orf92, KIAA1244, PPP1R33, dJ171N11.1 |
Location: |
Chr.6:138161690-138344663 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARFGEF3 | NM_020340.4 | NP_065073.3 | ||
| AB252196.1 | ||||
| BC137174.1 | ||||
| BF744369.1 | ||||
| BF943924.1 | ||||
| DT932721.1 |
Target Gene Details
Entrez Gene ID: | 59351 |
Gene Name: | prostate and breast cancer overexpressed 1 |
Gene Aliases: |
UC28, UROC28, dJ171N11.2 |
Location: |
Chr.6:138215986-138218490 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PBOV1 | NM_021635.2 | 1 | 354 | NP_067648.1 |
| AF189269.1 | 1 | 355 | AAG17117.1 | |
| AF189270.1 | 1 | 355 | AAG17118.1 | |
| BC069109.1 | 1 | 354 | AAH69109.1 |
More Information
Set Membership: |
|

Genomic Map