Assay Details
Target Gene Details
Entrez Gene ID: | 8553 |
Gene Name: | basic helix-loop-helix family member e40 |
Gene Aliases: |
BHLHB2, DEC1, HLHB2, SHARP-2, STRA13, Stra14 |
Location: |
Chr.3:4979412-4985181 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BHLHE40 | NM_003670.2 | 5 | 1569 | NP_003661.1 |
| AB004066.1 | 5 | 1458 | BAA21720.1 | |
| AK312509.1 | ||||
| BC082238.1 | 5 | 1443 | AAH82238.1 | |
| CR456921.1 | CAG33202.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005718 | Chr.3:4722315 - 5046200 on Build GRCh38 | Gain |
|
| nsv834606 | Chr.3:4835019 - 5057247 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map