Assay Details
Target Gene Details
Entrez Gene ID: | 55147 |
Gene Name: | RNA binding motif protein 23 |
Gene Aliases: |
CAPERbeta, PP239, RNPC4 |
Location: |
Chr.14:22900645-22919187 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
More Information
Additional Information:
For this assay, SNP(s) [rs72681969] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map