Assay Details
Target Gene Details
Entrez Gene ID: | 7352 |
Gene Name: | uncoupling protein 3 |
Gene Aliases: |
SLC25A9 |
Location: |
Chr.11:74000281-74009237 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UCP3 | NM_003356.3 | NP_003347.1 | ||
| NM_022803.2 | NP_073714.1 | |||
| AF001787.1 | AAC51369.1 | |||
| AF011449.1 | AAC51767.1 | |||
| AK092685.1 | ||||
| AK290630.1 | ||||
| AK291889.1 | ||||
| AK309873.1 | ||||
| BC008392.1 | AAH08392.1 | |||
| DA876282.1 | ||||
| KU178371.1 | ||||
| KU178372.1 | ||||
| U82818.1 | AAC51356.1 | |||
| U84763.1 | AAC51367.1 |
More Information
Set Membership: |
|

Genomic Map