Assay Details
Target Gene Details
Entrez Gene ID: | 81619 |
Gene Name: | tetraspanin 14 |
Gene Aliases: |
DC-TM4F2, TM4SF14 |
Location: |
Chr.10:80454271-80522635 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPAN14 | NM_001128309.1 | NP_001121781.1 | ||
| NM_030927.2 | NP_112189.2 | |||
| XM_006717992.3 | XP_006718055.1 | |||
| XM_011540219.2 | XP_011538521.1 | |||
| XM_011540220.2 | XP_011538522.1 | |||
| XM_011540221.2 | XP_011538523.1 | |||
| XM_011540222.2 | XP_011538524.1 | |||
| XM_017016707.1 | XP_016872196.1 | |||
| XM_017016708.1 | XP_016872197.1 | |||
| XM_017016709.1 | XP_016872198.1 | |||
| XM_017016710.1 | XP_016872199.1 | |||
| XM_017016711.1 | XP_016872200.1 | |||
| XM_017016712.1 | XP_016872201.1 | |||
| XM_017016713.1 | XP_016872202.1 | |||
| AF311903.1 | AAM94899.1 | |||
| AK074558.1 | BAC11059.1 | |||
| AK295321.1 | ||||
| AL136638.1 | CAB66573.1 | |||
| BC002920.2 | AAH02920.1 | |||
| BC093016.1 | AAH93016.1 | |||
| BI918996.1 | ||||
| DC300676.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469837 | Chr.10:80406994 - 80555534 on Build GRCh38 | Loss |
|
| nsv831923 | Chr.10:80323194 - 80509955 on Build GRCh38 | Loss |
|
| nsv831924 | Chr.10:80395862 - 80573439 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map