Assay Details
Target Gene Details
Entrez Gene ID: | 10420 |
Gene Name: | testis-specific kinase 2 |
Gene Aliases: |
- |
Location: |
Chr.1:45343883-45491342 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TESK2 | NM_001320800.1 | NP_001307729.1 | ||
| NM_007170.2 | NP_009101.2 | |||
| XM_005270355.3 | XP_005270412.1 | |||
| XM_006710287.2 | XP_006710350.1 | |||
| XM_011540497.1 | XP_011538799.1 | |||
| XM_017000047.1 | XP_016855536.1 | |||
| XM_017000048.1 | XP_016855537.1 | |||
| XM_017000049.1 | XP_016855538.1 | |||
| AB057597.1 | BAB62909.1 | |||
| AJ132545.1 | CAB41970.1 | |||
| AK027573.1 | ||||
| BC033085.1 | AAH33085.1 | |||
| BX648441.1 | ||||
| CD367940.1 |
Target Gene Details
Entrez Gene ID: | 114034 |
Gene Name: | target of EGR1, member 1 (nuclear) |
Gene Aliases: |
hCaf1z |
Location: |
Chr.1:45339457-45343978 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOE1 | NM_025077.3 | 8 | 2306 | NP_079353.3 |
| XM_005270412.3 | 8 | 2289 | XP_005270469.1 | |
| XM_005270413.4 | 8 | 2566 | XP_005270470.1 | |
| XM_011540569.2 | 7 | 2422 | XP_011538871.1 | |
| AK024011.1 | 8 | 1827 | BAB14774.1 | |
| AK093320.1 | 8 | 2101 | ||
| BC009364.2 | 8 | 1740 | AAH09364.1 | |
| BI495405.1 | 1 | 395 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2750837 | Chr.1:45330443 - 45442591 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map