Assay Details
Target Gene Details
Entrez Gene ID: | 6363 |
Gene Name: | C-C motif chemokine ligand 19 |
Gene Aliases: |
CKb11, ELC, MIP-3b, MIP3B, SCYA19 |
Location: |
Chr.9:34689570-34691277 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCL19 | NM_006274.2 | 2 | 235 | NP_006265.1 |
| AB000887.1 | 2 | 235 | BAA20383.1 | |
| AK311945.1 | 2 | 234 | ||
| AX772806.1 | 2 | 234 | CAE06252.1 | |
| BC027968.1 | 2 | 221 | AAH27968.1 | |
| CR456868.1 | 2 | 97 | CAG33149.1 | |
| U77180.1 | 2 | 97 | AAC50944.1 | |
| U88321.1 | 2 | 216 | AAC23905.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3620335 | Chr.9:33871811 - 34738198 on Build GRCh38 | Gain |
|
| nsv831544 | Chr.9:34650003 - 34818951 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map