Assay Details
Target Gene Details
Entrez Gene ID: | 23223 |
Gene Name: | ribosomal RNA processing 12 homolog |
Gene Aliases: |
KIAA0690 |
Location: |
Chr.10:97356701-97401370 on Build GRCh38 |
Assay Gene Location: | Within Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RRP12 | NM_001145114.1 | 32 | 4080 | NP_001138586.1 |
| NM_001284337.1 | 31 | 3963 | NP_001271266.1 | |
| NM_015179.3 | 34 | 4263 | NP_055994.2 | |
| XM_011539555.1 | 33 | 4132 | XP_011537857.1 | |
| AB014590.1 | 31 | 3876 | ||
| AK022496.1 | 34 | 4234 | ||
| AK056614.1 | 9 | 1413 | ||
| AK131283.1 | 28 | 3339 | ||
| AL832924.1 | 17 | 3109 | CAH10606.1 | |
| BC002621.2 | 7 | 843 | AAH02621.3 | |
| BC012745.2 | 34 | 4201 | AAH12745.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831952 | Chr.10:97238587 - 97386167 on Build GRCh38 | Loss |
|
| dgv119n27 | Chr.10:97355926 - 97414113 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map