Assay Details
Target Gene Details
Entrez Gene ID: | 51085 |
Gene Name: | MLX interacting protein like |
Gene Aliases: |
CHREBP, MIO, MONDOB, WBSCR14, WS-bHLH, bHLHd14 |
Location: |
Chr.7:73593194-73624543 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLXIPL | NM_032951.2 | NP_116569.1 | ||
| NM_032952.2 | NP_116570.1 | |||
| NM_032953.2 | NP_116571.1 | |||
| NM_032954.2 | NP_116572.1 | |||
| NR_134541.1 | ||||
| XM_011516277.1 | XP_011514579.1 | |||
| XM_011516278.1 | XP_011514580.1 | |||
| XM_011516279.1 | XP_011514581.1 | |||
| XM_011516281.2 | XP_011514583.1 | |||
| XM_017012263.1 | XP_016867752.1 | |||
| AF156603.1 | AAF68174.1 | |||
| AF245470.1 | AAK20935.1 | |||
| AF245471.1 | AAK20936.1 | |||
| AF245472.1 | AAK20937.1 | |||
| AF245473.1 | AAK20938.1 | |||
| AF245474.1 | AAK20939.1 | |||
| BC012925.1 | AAH12925.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023520 | Chr.7:73444580 - 73670500 on Build GRCh38 | Gain |
|
| nsv831028 | Chr.7:73556313 - 73816595 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map