Assay Details
Target Gene Details
Entrez Gene ID: | 3716 |
Gene Name: | Janus kinase 1 |
Gene Aliases: |
JAK1A, JAK1B, JTK3 |
Location: |
Chr.1:64833223-65067746 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAK1 | NM_001320923.1 | 26 | 4897 | NP_001307852.1 |
| NM_001321852.1 | 25 | 4900 | NP_001308781.1 | |
| NM_001321853.1 | 27 | 5159 | NP_001308782.1 | |
| NM_001321854.1 | 26 | 5075 | NP_001308783.1 | |
| NM_001321855.1 | 26 | 5078 | NP_001308784.1 | |
| NM_001321856.1 | 25 | 4813 | NP_001308785.1 | |
| NM_001321857.1 | 25 | 4991 | NP_001308786.1 | |
| NM_002227.3 | 25 | 4994 | NP_002218.2 | |
| XM_017001212.1 | 25 | 4838 | XP_016856701.1 | |
| BX647675.1 | 11 | 2598 |
More Information
Set Membership: |
|

Genomic Map