Assay Details
Target Gene Details
Entrez Gene ID: | 80218 |
Gene Name: | N(alpha)-acetyltransferase 50, NatE catalytic subunit |
Gene Aliases: |
MAK3, NAT13, NAT13P, NAT5, NAT5P, SAN |
Location: |
Chr.3:113716460-113746299 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA50 | NM_001308445.1 | 1 | 193 | NP_001295374.1 |
| NM_025146.3 | 1 | 193 | NP_079422.1 | |
| AK023090.1 | 1 | 108 | BAB14397.1 | |
| AK023256.1 | 1 | 143 | BAB14490.1 | |
| AK307184.1 | 1 | 132 | ||
| AL601126.1 | 1 | 91 | ||
| BC012731.2 | 1 | 132 | AAH12731.1 | |
| BM831426.1 | 1 | 167 | ||
| CR749314.1 | 1 | 88 | CAH18169.1 | |
| DA854531.1 | 1 | 193 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013506 | Chr.3:113171761 - 113962104 on Build GRCh38 | Gain |
|
| nsv829678 | Chr.3:113727330 - 113914206 on Build GRCh38 | Loss |
|
| nsv1013746 | Chr.3:113741315 - 113927810 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map