Assay Details
Target Gene Details
Entrez Gene ID: | 482 |
Gene Name: | ATPase Na+/K+ transporting subunit beta 2 |
Gene Aliases: |
AMOG |
Location: |
Chr.17:7646627-7657771 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP1B2 | NM_001303263.1 | 6 | 624 | NP_001290192.1 |
| NM_001678.4 | 7 | 1419 | NP_001669.3 | |
| AK290143.1 | 7 | 1161 | ||
| BC126175.1 | 7 | 1127 | ||
| BG708115.1 | 6 | 615 | ||
| M81181.1 | 7 | 1197 | AAA51805.1 | |
| U45945.1 | 7 | 826 | AAC50873.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv457659 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map