Assay Details
Target Gene Details
Entrez Gene ID: | 23125 |
Gene Name: | calmodulin binding transcription activator 2 |
Gene Aliases: |
- |
Location: |
Chr.17:4967992-4988297 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAMTA2 | NM_001171166.1 | 21 | 4329 | NP_001164637.1 |
| NM_001171167.1 | 23 | 4787 | NP_001164638.1 | |
| NM_001171168.1 | 22 | 4717 | NP_001164639.1 | |
| NM_015099.3 | 23 | 4445 | NP_055914.2 | |
| XM_006721478.3 | 23 | 5092 | XP_006721541.1 | |
| XM_006721481.3 | 22 | 5005 | XP_006721544.1 | |
| XM_006721482.3 | 21 | 4138 | XP_006721545.1 | |
| XM_011523746.2 | 23 | 4452 | XP_011522048.1 | |
| XM_011523747.2 | 23 | 4436 | XP_011522049.1 | |
| XM_011523748.2 | 22 | 4430 | XP_011522050.1 | |
| XM_011523749.2 | 22 | 4414 | XP_011522051.1 | |
| AB020716.1 | 23 | 4406 | ||
| AL831849.1 | 22 | 4717 | CAD38553.1 | |
| AL833974.1 | 21 | 4314 | CAD38818.2 | |
| BC010050.2 | 14 | 2837 | AAH10050.2 | |
| BC016163.1 | 10 | 2064 | AAH16163.1 | |
| BC032381.1 | 7 | 1460 | AAH32381.1 | |
| BC142695.1 | 23 | 4436 | ||
| BC167148.1 | 23 | 4406 | ||
| BC167149.1 | 23 | 4406 | ||
| BC167151.1 | 23 | 4406 | ||
| BC167160.1 | 23 | 4406 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833350 | Chr.17:4884713 - 5060776 on Build GRCh38 | Loss |
|
| nsv833349 | Chr.17:4853631 - 4991300 on Build GRCh38 | Loss |
|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map