Assay Details
Target Gene Details
Entrez Gene ID: | 5892 |
Gene Name: | RAD51 paralog D |
Gene Aliases: |
BROVCA4, R51H3, RAD51L3, TRAD |
Location: |
Chr.17:35099792-35119869 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD51D | NM_001142571.1 | 10 | 1376 | NP_001136043.1 |
| NM_002878.3 | 10 | 1316 | NP_002869.3 | |
| NM_133629.2 | 7 | 980 | NP_598332.1 | |
| NR_037711.1 | 9 | 1197 | ||
| NR_037712.1 | 8 | 1062 | ||
| AB013341.1 | 10 | 1166 | BAA25914.1 | |
| AB016223.1 | 9 | 1047 | BAA31747.1 | |
| AB016224.1 | 8 | 912 | BAA31748.1 | |
| AB016225.1 | 7 | 830 | BAA31749.1 | |
| AF034956.1 | 9 | 1067 | AAC39719.1 | |
| AK296241.1 | 10 | 1320 | ||
| AK301249.1 | 10 | 1382 | ||
| AL117459.1 | 6 | 597 | CAB55937.1 | |
| BC002723.2 | 9 | 1081 | AAH02723.1 | |
| BC014422.1 | 10 | 1195 | AAH14422.1 | |
| BX647297.1 | 10 | 1263 | ||
| Y15572.1 | 10 | 1179 | CAA75681.1 |
Target Gene Details
Entrez Gene ID: | 100529207 |
Gene Name: | RAD51L3-RFFL readthrough |
Gene Aliases: |
- |
Location: |
Chr.17:35009112-35121522 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD51L3-RFFL | NR_037714.1 | |||
| AK097811.1 |
More Information
Set Membership: |
|

Genomic Map