Assay Details
Target Gene Details
Entrez Gene ID: | 55973 |
Gene Name: | B-cell receptor-associated protein 29 |
Gene Aliases: |
BAP29 |
Location: |
Chr.7:107579977-107623317 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCAP29 | NM_001008405.2 | NP_001008405.1 | ||
| NM_018844.3 | NP_061332.2 | |||
| NR_027830.1 | ||||
| XM_006716051.2 | XP_006716114.1 | |||
| XM_011516406.2 | XP_011514708.1 | |||
| XM_011516407.1 | XP_011514709.1 | |||
| XM_011516408.2 | XP_011514710.1 | |||
| XM_011516409.2 | XP_011514711.1 | |||
| XM_017012436.1 | XP_016867925.1 | |||
| XM_017012437.1 | XP_016867926.1 | |||
| XM_017012438.1 | XP_016867927.1 | |||
| AA826200.1 | ||||
| AF126020.1 | AAF17230.1 | |||
| AK289401.1 | ||||
| AK294824.1 | ||||
| AK298594.1 | ||||
| AW978976.1 | ||||
| BC008478.1 | AAH08478.1 | |||
| BM023571.1 | ||||
| BP261060.1 | ||||
| BT006981.1 | AAP35627.1 | |||
| BX457671.2 | ||||
| BX647653.1 | ||||
| CK820004.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv608074 | Chr.7:107538686 - 107620411 on Build GRCh38 | Loss |
|
| esv3614462 | Chr.7:107566581 - 107659648 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map