Assay Details
Target Gene Details
Entrez Gene ID: | 115209 |
Gene Name: | OMA1 zinc metallopeptidase |
Gene Aliases: |
2010001O09Rik, DAB1, MPRP-1, YKR087C, ZMPOMA1, peptidase |
Location: |
Chr.1:58480719-58546799 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OMA1 | NM_145243.4 | 9 | 1679 | NP_660286.1 |
| AB048348.1 | 9 | 1662 | BAC79381.1 | |
| AK091101.1 | BAC03583.1 | |||
| BC012915.1 | 9 | 1660 | AAH12915.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829948 | Chr.1:58338009 - 58505708 on Build GRCh38 | Gain |
|
| nsv1003930 | Chr.1:58464772 - 58691366 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map