Assay Details
Target Gene Details
Entrez Gene ID: | 126382 |
Gene Name: | nuclear receptor 2C2 associated protein |
Gene Aliases: |
TRA16 |
Location: |
Chr.19:19201409-19203453 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NR2C2AP | NM_001300945.1 | NP_001287874.1 | ||
| NM_176880.5 | NP_795361.1 | |||
| XM_011527688.2 | 1 | 25 | XP_011525990.1 | |
| BC057837.1 | AAH57837.1 | |||
| CF993811.1 | ||||
| HY088736.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1151217 | Chr.19:18627369 - 19284914 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map