Assay Details
Target Gene Details
Entrez Gene ID: | 4015 |
Gene Name: | lysyl oxidase |
Gene Aliases: |
- |
Location: |
Chr.5:122063195-122078501 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOX | NM_001178102.2 | NP_001171573.1 | ||
| NM_001317073.1 | NP_001304002.1 | |||
| NM_002317.6 | NP_002308.2 | |||
| AF039291.1 | AAD02130.1 | |||
| AK297709.1 | ||||
| AK298781.1 | ||||
| AK307559.1 | ||||
| AK307732.1 | ||||
| AK311354.1 | ||||
| AK312269.1 | ||||
| AK315873.1 | ||||
| AK316309.1 | ||||
| BC074820.2 | AAH74820.1 | |||
| BC074872.2 | AAH74872.1 | |||
| BC089436.1 | AAH89436.1 | |||
| CN265862.1 | ||||
| DA720490.1 | ||||
| M94054.1 | AAA59525.1 | |||
| S45875.1 | AAB23549.1 | |||
| S78694.1 | AAB21243.1 |
Target Gene Details
Entrez Gene ID: | 153443 |
Gene Name: | serum response factor binding protein 1 |
Gene Aliases: |
BUD22, P49, Rlb1, STRAP, p49/STRAP |
Location: |
Chr.5:121961917-122075624 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRFBP1 | XM_017009111.1 | 8 | 1344 | XP_016864600.1 |
More Information
Set Membership: |
|

Genomic Map