Assay Details
Target Gene Details
Entrez Gene ID: | 56938 |
Gene Name: | aryl hydrocarbon receptor nuclear translocator like 2 |
Gene Aliases: |
BMAL2, CLIF, MOP9, PASD9, bHLHe6 |
Location: |
Chr.12:27332854-27425813 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARNTL2 | NM_001248002.1 | 9 | 1151 | NP_001234931.1 |
| NM_001248003.1 | 8 | 1082 | NP_001234932.1 | |
| NM_001248004.1 | 8 | 1049 | NP_001234933.1 | |
| NM_001248005.1 | 8 | 1082 | NP_001234934.1 | |
| NM_020183.4 | 10 | 1193 | NP_064568.3 | |
| XM_006719112.3 | 9 | 1152 | XP_006719175.1 | |
| XM_006719114.3 | 9 | 883 | XP_006719177.1 | |
| XM_011520766.2 | 10 | 1194 | XP_011519068.1 | |
| XM_011520768.2 | 9 | 1102 | XP_011519070.1 | |
| XM_011520769.2 | 9 | 1092 | XP_011519071.1 | |
| XM_011520770.2 | 10 | 1012 | XP_011519072.1 | |
| XM_011520771.2 | 10 | 978 | XP_011519073.1 | |
| XM_017019666.1 | 7 | 923 | XP_016875155.1 | |
| XM_017019667.1 | 10 | 1210 | XP_016875156.1 | |
| XM_017019668.1 | 10 | 1007 | XP_016875157.1 | |
| XM_017019669.1 | 9 | 854 | XP_016875158.1 | |
| AB039921.1 | 7 | 725 | BAB01485.1 | |
| AF231338.1 | 8 | 878 | AAF71306.1 | |
| AF231339.1 | 7 | 839 | AAF71307.1 | |
| AF246960.1 | 10 | 992 | AAL50339.1 | |
| AF246961.1 | 9 | 950 | AAL50340.1 | |
| AF246962.1 | 8 | 881 | AAL50341.1 | |
| AF246963.1 | 8 | 848 | AAL50342.1 | |
| AK296706.1 | 8 | 1016 | ||
| BC000172.3 | 7 | 800 | AAH00172.3 | |
| BC125061.1 | 8 | 843 | ||
| BC125062.1 | 8 | 881 |
Target Gene Details
Entrez Gene ID: | 101928646 |
Gene Name: | ARNTL2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.12:27389789-27446634 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARNTL2-AS1 | NR_109975.1 | |||
| BU568793.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1406n100 | Chr.12:27128236 - 27640822 on Build GRCh38 | Gain |
|
| dgv2417n54 | Chr.12:27068022 - 27638988 on Build GRCh38 | Gain |
|
| esv3628933 | Chr.12:27275994 - 27403117 on Build GRCh38 | Gain |
|
| nsv436825 | Chr.12:27135971 - 27642579 on Build GRCh38 | Insertion |
|
| esv2745664 | Chr.12:27017516 - 27820117 on Build GRCh38 | Deletion |
|
| nsv832357 | Chr.12:27399918 - 27439033 on Build GRCh38 | Gain |
|
| dgv271e199 | Chr.12:27089700 - 27631451 on Build GRCh38 | Deletion |
|
| nsv521872 | Chr.12:27114021 - 27635541 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map