Assay Details
Target Gene Details
Entrez Gene ID: | 22930 |
Gene Name: | RAB3 GTPase activating protein catalytic subunit 1 |
Gene Aliases: |
P130, RAB3GAP, RAB3GAP130, WARBM1 |
Location: |
Chr.2:135052265-135176667 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB3GAP1 | NM_001172435.1 | NP_001165906.1 | ||
| NM_012233.2 | NP_036365.1 | |||
| XM_011510823.2 | XP_011509125.1 | |||
| XM_011510825.2 | XP_011509127.1 | |||
| AB449877.1 | ||||
| AK022494.1 | ||||
| BC146809.1 | ||||
| BP872264.1 | ||||
| D31886.1 | BAA06684.1 | |||
| DA333844.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3592568 | Chr.2:134994189 - 135317860 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map