Assay Details
Target Gene Details
Entrez Gene ID: | 51660 |
Gene Name: | mitochondrial pyruvate carrier 1 |
Gene Aliases: |
BRP44L, CGI-129, MPYCD, dJ68L15.3 |
Location: |
Chr.6:166364919-166383013 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPC1 | NM_001270879.1 | 6 | 1130 | NP_001257808.1 |
| NM_016098.3 | 5 | 914 | NP_057182.1 | |
| NR_073093.1 | 6 | 1143 | ||
| XM_011535894.1 | 5 | 1913 | XP_011534196.1 | |
| XM_011535895.1 | 4 | 1697 | XP_011534197.1 | |
| AF125101.1 | 5 | 899 | AAD39918.1 | |
| AF151887.1 | 5 | 896 | AAD34124.1 | |
| AF275811.1 | 3 | 621 | AAG23822.1 | |
| AL556427.3 | 5 | 833 | ||
| AL561589.3 | ||||
| BC000810.1 | 5 | 824 | AAH00810.1 |
More Information
Set Membership: |
|

Genomic Map