Assay Details
Target Gene Details
Entrez Gene ID: | 4150 |
Gene Name: | MYC associated zinc finger protein |
Gene Aliases: |
PUR1, Pur-1, SAF-1, SAF-2, SAF-3, ZF87, ZNF801, Zif87 |
Location: |
Chr.16:29806096-29811183 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAZ | NM_001042539.2 | NP_001036004.1 | ||
| NM_001276275.1 | NP_001263204.1 | |||
| NM_001276276.1 | NP_001263205.1 | |||
| NM_002383.3 | NP_002374.2 | |||
| NR_074080.1 | ||||
| XM_006721047.3 | XP_006721110.1 | |||
| AB209061.1 | 1 | 2063 | BAD92298.1 | |
| AF489858.1 | AAN03800.1 | |||
| AK074720.1 | ||||
| BC041629.1 | AAH41629.1 | |||
| BI198991.1 | ||||
| BX362240.2 | ||||
| D85131.1 | BAA12728.1 | |||
| EF059746.1 | ||||
| FJ532357.1 | ||||
| L01420.1 | AAL31317.1 | |||
| M93339.1 | ||||
| M94046.1 | ||||
| U33819.1 | AAB04121.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833186 | Chr.16:29719552 - 29863674 on Build GRCh38 | Loss |
|
| nsv953800 | Chr.16:29804780 - 29816179 on Build GRCh38 | Deletion |
|
| nsv827611 | Chr.16:29808186 - 29830422 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map