Assay Details
Target Gene Details
Entrez Gene ID: | 164284 |
Gene Name: | APC down-regulated 1 like |
Gene Aliases: |
- |
Location: |
Chr.20:58459101-58514938 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APCDD1L | NM_001304787.1 | 5 | 1596 | NP_001291716.1 |
| NM_153360.2 | 4 | 1425 | NP_699191.1 | |
| AK074647.1 | 4 | 1380 | BAC11111.1 | |
| AK293299.1 | 5 | 1555 | ||
| BC101758.1 | 4 | 1277 | AAI01759.1 | |
| BC101760.1 | 4 | 1277 | AAI01761.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3428 | Chr.20:58451051 - 58496125 on Build GRCh38 | Deletion |
|
| nsv834019 | Chr.20:58397598 - 58607087 on Build GRCh38 | Loss |
|

Genomic Map