Assay Details
Target Gene Details
Entrez Gene ID: | 55924 |
Gene Name: | family with sequence similarity 212 member B |
Gene Aliases: |
C1orf183 |
Location: |
Chr.1:111722064-111755797 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM212B | NM_019099.4 | 2 | 1095 | NP_061972.1 |
| NM_198926.2 | 2 | 953 | NP_945120.1 | |
| XM_011541783.2 | 4 | 1191 | XP_011540085.1 | |
| AK055667.1 | 2 | 1071 | ||
| AK293989.1 | 2 | 980 | ||
| AK295012.1 | ||||
| AL137198.1 | 2 | 1002 | CAB69908.1 | |
| AL137199.1 | 2 | 953 | CAB69909.1 | |
| BC031558.1 | 2 | 1050 | AAH31558.1 |

Genomic Map