Assay Details
Target Gene Details
Entrez Gene ID: | 6520 |
Gene Name: | solute carrier family 3 member 2 |
Gene Aliases: |
4F2, 4F2HC, 4T2HC, CD98, CD98HC, MDU1, NACAE |
Location: |
Chr.11:62856012-62888883 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC3A2 | NM_001012662.2 | 1 | 158 | NP_001012680.1 |
| NM_001012664.2 | 1 | 158 | NP_001012682.1 | |
| NM_002394.5 | 1 | 158 | NP_002385.3 | |
| AK025584.1 | 1 | 59 | ||
| AU143131.1 | 1 | 121 | ||
| BC001061.2 | 1 | 68 | AAH01061.2 | |
| BC003000.1 | AAH03000.2 | |||
| BE018712.1 | 1 | 113 | ||
| BU556953.1 | 1 | 82 | ||
| BX362778.2 | 1 | 125 | ||
| CB138799.1 | ||||
| CN302816.1 | 1 | 138 | ||
| DC395235.1 | 1 | 158 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1159903 | Chr.11:62818434 - 62885307 on Build GRCh38 | Duplication |
|
| nsv509416 | Chr.11:62779642 - 62876986 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map