Assay Details
Target Gene Details
Entrez Gene ID: | 85320 |
Gene Name: | ATP binding cassette subfamily C member 11 |
Gene Aliases: |
EWWD, MRP8, WW |
Location: |
Chr.16:48164842-48249973 on Build GRCh38 |
Assay Gene Location: | Within Exon 37 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCC11 | NM_032583.3 | 30 | 4369 | NP_115972.2 |
| NM_033151.3 | 29 | 4655 | NP_149163.2 | |
| NM_145186.2 | 29 | 4255 | NP_660187.1 | |
| XM_011523397.1 | 26 | 3875 | XP_011521699.1 | |
| XM_011523398.2 | 18 | 2590 | XP_011521700.1 | |
| XM_017023795.1 | 30 | 6983 | XP_016879284.1 | |
| XM_017023796.1 | 31 | 4456 | XP_016879285.1 | |
| XM_017023797.1 | 31 | 4584 | XP_016879286.1 | |
| XM_017023798.1 | 30 | 4410 | XP_016879287.1 | |
| XM_017023799.1 | 30 | 4392 | XP_016879288.1 | |
| XM_017023800.1 | 30 | 4383 | XP_016879289.1 | |
| XM_017023801.1 | 29 | 6875 | XP_016879290.1 | |
| XM_017023802.1 | 24 | 3730 | XP_016879291.1 | |
| AF352582.1 | 30 | 4404 | AAK19755.1 | |
| AF367202.1 | 30 | 4383 | AAK58869.1 | |
| AF411579.1 | 29 | 4269 | AAL99902.1 | |
| AL117406.1 | 11 | 1605 | ||
| AY040219.1 | 29 | 4655 | AAK76739.1 |
More Information
Set Membership: |
|

Genomic Map