Assay Details
Target Gene Details
Entrez Gene ID: | 221037 |
Gene Name: | jumonji domain containing 1C |
Gene Aliases: |
TRIP-8, TRIP8 |
Location: |
Chr.10:63167221-63522075 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JMJD1C | NM_001282948.1 | 18 | 6854 | NP_001269877.1 |
| NM_001318153.1 | 18 | 6858 | NP_001305082.1 | |
| NM_001318154.1 | 19 | 6796 | NP_001305083.1 | |
| NM_001322252.1 | 18 | 6851 | NP_001309181.1 | |
| NM_001322254.1 | 17 | 6748 | NP_001309183.1 | |
| NM_001322258.1 | 18 | 6690 | NP_001309187.1 | |
| NM_032776.2 | 19 | 6965 | NP_116165.1 | |
| NR_134512.1 | 15 | 6378 | ||
| XM_011539503.1 | 18 | 6458 | XP_011537805.1 | |
| XM_017015897.1 | 18 | 6453 | XP_016871386.1 | |
| XM_017015898.1 | 19 | 6646 | XP_016871387.1 | |
| XM_017015899.1 | 19 | 6546 | XP_016871388.1 | |
| XM_017015900.1 | 19 | 6950 | XP_016871389.1 | |
| XM_017015901.1 | 18 | 6493 | XP_016871390.1 | |
| XM_017015902.1 | 18 | 6754 | XP_016871391.1 | |
| XM_017015903.1 | 17 | 6302 | XP_016871392.1 | |
| AB037801.1 | 10 | 2802 | BAA92618.1 | |
| AK024991.1 | 8 | 2265 | ||
| AL831917.1 | 12 | 5493 | CAD38578.1 | |
| BC143722.1 | 18 | 6697 | ||
| BX537954.1 | 15 | 6341 | CAD97921.1 | |
| DR762886.1 | 2 | 252 | ||
| EF068222.1 | 21 | 6922 | ||
| L40411.1 | AAC41741.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2677353 | Chr.10:63130283 - 63501631 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map