Assay Details
Target Gene Details
Entrez Gene ID: | 347088 |
Gene Name: | adhesion G protein-coupled receptor D2 |
Gene Aliases: |
GPR144, PGR24 |
Location: |
Chr.9:124451087-124477795 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRD2 | XM_006717348.3 | 3 | 262 | XP_006717411.2 |
| AY278562.1 | 3 | 205 | AAP35064.1 | |
| EF107715.1 | ||||
| EF107717.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv615290 | Chr.9:124355067 - 124455979 on Build GRCh38 | Loss |
|
| esv3621722 | Chr.9:124378965 - 124497390 on Build GRCh38 | Gain |
|
| nsv950941 | Chr.9:124444622 - 124456821 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map