Assay Details
Target Gene Details
Entrez Gene ID: | 162387 |
Gene Name: | major facilitator superfamily domain containing 6 like |
Gene Aliases: |
- |
Location: |
Chr.17:8797110-8799349 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFSD6L | NM_152599.3 | 1 | 1305 | NP_689812.3 |
| AK093092.1 | 1 | 1258 | BAC04050.1 | |
| AY129026.1 | 1 | 1265 | AAM98769.1 | |
| BC040487.1 | 1 | 1224 | AAH40487.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518675 | Chr.17:8696386 - 8818290 on Build GRCh38 | Gain |
|
| nsv953828 | Chr.17:8796283 - 8806582 on Build GRCh38 | Deletion |
|
| nsv1969 | Chr.17:8780681 - 8825867 on Build GRCh38 | Deletion |
|

Genomic Map