Assay Details
Target Gene Details
Entrez Gene ID: | 2921 |
Gene Name: | C-X-C motif chemokine ligand 3 |
Gene Aliases: |
CINC-2b, GRO3, GROg, MIP-2b, MIP2B, SCYB3 |
Location: |
Chr.4:74036589-74038773 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCL3 | NM_002090.2 | 4 | 1032 | NP_002081.2 |
| AW079760.1 | 1 | 135 | ||
| BC016308.1 | 4 | 952 | AAH16308.1 | |
| BC065743.1 | 4 | 952 | AAH65743.1 | |
| M36821.1 | 4 | 944 | AAA63184.1 | |
| X53800.1 | 4 | 904 | CAA37809.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006029 | Chr.4:73928205 - 74120947 on Build GRCh38 | Gain |
|
| esv3575747 | Chr.4:73914851 - 74124725 on Build GRCh38 | Gain |
|
| nsv525980 | Chr.4:73865809 - 74556070 on Build GRCh38 | Gain |
|
| nsv1004768 | Chr.4:73908655 - 74243862 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map