Assay Details
Target Gene Details
Entrez Gene ID: | 84824 |
Gene Name: | Fc receptor like A |
Gene Aliases: |
FCRL, FCRL1, FCRLM1, FCRLX, FCRLb, FCRLc1, FCRLc2, FCRLd, FCRLe, FCRX, FREB |
Location: |
Chr.1:161706972-161714352 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428257 | Chr.1:161436525 - 161898473 on Build GRCh38 | Loss |
|
| nsv464184 | Chr.1:161677017 - 161915562 on Build GRCh38 | Gain |
|
| esv2758972 | Chr.1:161374465 - 161711762 on Build GRCh38 | Gain+Loss |
|
| esv33393 | Chr.1:161508891 - 162110537 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map