Assay Details
Target Gene Details
Entrez Gene ID: | 7804 |
Gene Name: | LDL receptor related protein 8 |
Gene Aliases: |
APOER2, HSZ75190, LRP-8, MCI1 |
Location: |
Chr.1:53242364-53328149 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011750 | Chr.1:53226981 - 53261502 on Build GRCh38 | Gain |
|
| esv2422213 | Chr.1:53179891 - 53258767 on Build GRCh38 | Duplication |
|
| nsv829871 | Chr.1:53200871 - 53383048 on Build GRCh38 | Loss |
|
| nsv1001535 | Chr.1:52620368 - 53346539 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map