Assay Details
Target Gene Details
Entrez Gene ID: | 151126 |
Gene Name: | zinc finger protein 385B |
Gene Aliases: |
ZNF533 |
Location: |
Chr.2:179441982-179864550 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 22 |
More Information
Set Membership: |
|

Genomic Map