Assay Details
Target Gene Details
Entrez Gene ID: | 139285 |
Gene Name: | APC membrane recruitment protein 1 |
Gene Aliases: |
FAM123B, OSCS, WTX |
Location: |
Chr.X:64185117-64205744 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMER1 | NM_152424.3 | 2 | 8169 | NP_689637.3 |
| AK097146.1 | 3 | 3414 | BAC04964.1 | |
| AL833759.1 | 1 | 1556 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521560 | Chr.X:64063724 - 64245972 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map