Assay Details
Target Gene Details
Entrez Gene ID: | 116337 |
Gene Name: | pannexin 3 |
Gene Aliases: |
PX3 |
Location: |
Chr.11:124611557-124620356 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PANX3 | NM_052959.2 | NP_443191.1 | ||
| AF406650.1 | AAK95655.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045156 | Chr.11:124565356 - 124705852 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map