Assay Details
Target Gene Details
Entrez Gene ID: | 55679 |
Gene Name: | LIM zinc finger domain containing 2 |
Gene Aliases: |
LGMD2W, PINCH-2, PINCH2 |
Location: |
Chr.2:127638421-127681786 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIMS2 | NM_001136037.2 | NP_001129509.2 | ||
| NM_001161403.1 | NP_001154875.1 | |||
| NM_001161404.1 | NP_001154876.1 | |||
| NM_001256542.1 | NP_001243471.1 | |||
| NM_017980.4 | NP_060450.2 | |||
| XM_005263710.2 | XP_005263767.1 | |||
| XM_006712627.3 | XP_006712690.1 | |||
| XM_011511453.1 | XP_011509755.1 | |||
| XM_017004469.1 | XP_016859958.1 | |||
| AF484961.1 | AAM97589.1 | |||
| AF520987.1 | AAM77350.1 | |||
| AF527764.1 | AAQ09011.1 | |||
| AF527765.1 | AAQ09012.1 | |||
| AF527766.1 | AAQ09013.1 | |||
| AF527767.1 | AAQ09014.1 | |||
| AF527768.1 | AAQ09015.1 | |||
| AF527769.1 | AAQ09016.1 | |||
| AF527770.1 | AAQ09017.1 | |||
| AK000906.1 | BAA91419.1 | |||
| AK022470.1 | BAB14047.1 | |||
| AK055363.1 | ||||
| AK091652.1 | ||||
| AK093692.1 | 2 | 378 | ||
| AK094954.1 | 7 | 871 | ||
| AK095790.1 | ||||
| AK123014.1 | ||||
| AK297645.1 | ||||
| BC065816.1 | AAH65816.1 | |||
| BX458594.2 | ||||
| DA848179.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517532 | Chr.2:127630362 - 127647285 on Build GRCh38 | Loss |
|
| dgv4068n100 | Chr.2:127537744 - 127672201 on Build GRCh38 | Gain |
|
| esv3893304 | Chr.2:127634972 - 127643416 on Build GRCh38 | Loss |
|
| nsv508864 | Chr.2:127585891 - 127684658 on Build GRCh38 | Insertion |
|
| nsv953155 | Chr.2:127617926 - 127665326 on Build GRCh38 | Deletion |
|
| nsv518085 | Chr.2:127576838 - 127647285 on Build GRCh38 | Loss |
|
| nsv470482 | Chr.2:127593233 - 127707973 on Build GRCh38 | Loss |
|
| nsv583037 | Chr.2:127576895 - 127643516 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map