Assay Details
Target Gene Details
Entrez Gene ID: | 83538 |
Gene Name: | tetratricopeptide repeat domain 25 |
Gene Aliases: |
- |
Location: |
Chr.17:41930605-41966517 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC25 | NM_031421.3 | 3 | 392 | NP_113609.1 |
| NR_110662.1 | 3 | 392 | ||
| XM_011525323.2 | XP_011523625.1 | |||
| XM_011525324.2 | 3 | 359 | XP_011523626.1 | |
| AK055498.1 | 3 | 337 | BAB70936.1 | |
| AL136760.1 | 3 | 359 | CAB66694.1 | |
| BC025390.1 | 3 | 400 | AAH25390.1 | |
| BC067297.1 | 3 | 351 | AAH67297.1 | |
| CF593846.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509658 | Chr.17:41927491 - 42002888 on Build GRCh38 | Insertion |
|
| nsv2056 | Chr.17:41923301 - 41975161 on Build GRCh38 | Insertion |
|
| nsv833447 | Chr.17:41794279 - 41955275 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map