Assay Details
Target Gene Details
Entrez Gene ID: | 51428 |
Gene Name: | DEAD-box helicase 41 |
Gene Aliases: |
ABS, MPLPF |
Location: |
Chr.5:177511577-177517326 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX41 | NM_001321732.1 | NP_001308661.1 | ||
| NM_001321830.1 | NP_001308759.1 | |||
| NM_016222.3 | NP_057306.2 | |||
| AF195417.1 | AAF04150.1 | |||
| AK001255.1 | BAA91585.1 | |||
| AK026117.1 | ||||
| AK027768.1 | BAB55355.1 | |||
| AK091545.1 | ||||
| AK091774.1 | ||||
| AK092585.1 | ||||
| AK222598.1 | BAD96318.1 | |||
| AK315491.1 | ||||
| AL137455.1 | CAB70746.1 | |||
| BC015476.1 | AAH15476.1 | |||
| BX641072.1 | CAE46035.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5147 | Chr.5:177511195 - 177556017 on Build GRCh38 | Deletion |
|
| nsv1161311 | Chr.5:177305104 - 177546474 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map