Assay Details
Target Gene Details
Entrez Gene ID: | 57466 |
Gene Name: | SR-related CTD associated factor 4 |
Gene Aliases: |
SFRS15, SRA4 |
Location: |
Chr.21:31671000-31732118 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCAF4 | NM_001145444.1 | 19 | 3871 | NP_001138916.1 |
| NM_001145445.1 | 20 | 3850 | NP_001138917.1 | |
| NM_020706.2 | 20 | 3916 | NP_065757.1 | |
| XM_005261017.2 | 19 | 3720 | XP_005261074.1 | |
| XM_006724035.2 | 20 | 3885 | XP_006724098.1 | |
| XM_006724036.2 | 20 | 3819 | XP_006724099.1 | |
| XM_017028415.1 | 20 | 3874 | XP_016883904.1 | |
| XM_017028416.1 | 20 | 3693 | XP_016883905.1 | |
| XM_017028417.1 | 19 | 3654 | XP_016883906.1 | |
| AB032998.1 | 14 | 2838 | BAA86486.1 | |
| AF023142.1 | 20 | 3515 | AAD09327.1 | |
| AK025748.1 | 15 | 2869 | ||
| AK027017.1 | 6 | 1661 | ||
| AK308406.1 | 19 | 3722 | ||
| AL834304.1 | 20 | 3630 | CAD38974.2 | |
| BC014921.1 | 20 | 3893 | ||
| BC043353.1 | 20 | 3873 | ||
| BC052286.1 | 20 | 3747 | ||
| BC064990.1 | 20 | 3747 | AAH64990.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1067151 | Chr.21:31374681 - 32180843 on Build GRCh38 | Gain |
|
| nsv519899 | Chr.21:31509375 - 31691041 on Build GRCh38 | Gain |
|
| esv3575385 | Chr.21:31000408 - 31807461 on Build GRCh38 | Gain |
|
| nsv834079 | Chr.21:31604633 - 31781547 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map