Assay Details
Target Gene Details
Entrez Gene ID: | 50650 |
Gene Name: | Rho guanine nucleotide exchange factor 3 |
Gene Aliases: |
GEF3, STA3, XPLN |
Location: |
Chr.3:56727418-57079308 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF3 | NM_001128615.1 | NP_001122087.1 | ||
| XM_005265186.4 | XP_005265243.1 | |||
| XM_005265187.3 | XP_005265244.1 | |||
| XM_011533765.1 | XP_011532067.1 | |||
| XM_011533766.2 | XP_011532068.1 | |||
| XM_017006502.1 | XP_016861991.1 | |||
| XM_017006503.1 | XP_016861992.1 | |||
| AF433662.1 | AAP97313.1 | |||
| AK302493.1 | ||||
| DC334772.1 |
Target Gene Details
Entrez Gene ID: | 353324 |
Gene Name: | spermatogenesis associated 12 |
Gene Aliases: |
SRG5 |
Location: |
Chr.3:57060132-57076068 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPATA12 | NM_181727.1 | 2 | 965 | NP_859078.1 |
| XM_011533676.2 | 4 | 1511 | XP_011531978.1 | |
| AY221117.1 | 2 | 965 | AAP50852.1 | |
| BC033337.1 | 2 | 740 | ||
| BC126274.1 | 2 | 894 | ||
| BC136498.1 | 2 | 894 |
More Information
Set Membership: |
|

Genomic Map