Assay Details
Target Gene Details
Entrez Gene ID: | 84678 |
Gene Name: | lysine demethylase 2B |
Gene Aliases: |
CXXC2, FBXL10, Fbl10, JHDM1B, PCCX2 |
Location: |
Chr.12:121402289-121582268 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM2B | NM_001005366.1 | NP_001005366.1 | ||
| NM_032590.4 | NP_115979.3 | |||
| XM_005253955.3 | XP_005254012.1 | |||
| XM_005253956.3 | XP_005254013.1 | |||
| XM_005253961.4 | XP_005254018.1 | |||
| XM_011538867.2 | XP_011537169.1 | |||
| XM_011538868.2 | XP_011537170.1 | |||
| XM_011538869.2 | XP_011537171.1 | |||
| XM_011538875.2 | XP_011537177.1 | |||
| AB031230.1 | BAA97672.1 | |||
| AJ459424.1 | CAD30700.1 | |||
| AK027440.1 | BAB55112.1 | |||
| AK027692.1 | BAB55301.1 | |||
| AK127328.1 | ||||
| AK299827.1 | ||||
| AK316470.1 | ||||
| AL133572.1 | CAB63721.2 | |||
| BC008735.2 | AAH08735.2 | |||
| BC115379.1 | AAI15380.1 | |||
| BC115380.1 | AAI15381.1 |
Target Gene Details
Entrez Gene ID: | 102465665 |
Gene Name: | microRNA 7107 |
Gene Aliases: |
hsa-mir-7107 |
Location: |
Chr.12:121444273-121444352 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR7107 | NR_106957.1 |
More Information
Set Membership: |
|

Genomic Map