Assay Details
Target Gene Details
Entrez Gene ID: | 2558 |
Gene Name: | gamma-aminobutyric acid type A receptor alpha5 subunit |
Gene Aliases: |
- |
Location: |
Chr.15:26866719-26949210 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GABRA5 | NM_000810.3 | 1 | 335 | NP_000801.1 |
| A28104.1 | 1 | 101 | CAA01920.1 | |
| AK290453.1 | ||||
| DA157081.1 | 1 | 335 | ||
| L08485.1 | 1 | 109 | AAA58490.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2668795 | Chr.15:26828644 - 27745218 on Build GRCh38 | Deletion |
|
| esv23657 | Chr.15:26867088 - 26868198 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map