Assay Details
Target Gene Details
Entrez Gene ID: | 55572 |
Gene Name: | FAD dependent oxidoreductase domain containing 1 |
Gene Aliases: |
FP634, H17 |
Location: |
Chr.11:126269040-126278132 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXRED1 | NM_017547.3 | 11 | 1663 | NP_060017.1 |
| NR_037647.1 | 10 | 1442 | ||
| NR_037648.1 | 11 | 1682 | ||
| XM_006718879.3 | 10 | 1376 | XP_006718942.1 | |
| XM_006718881.3 | 9 | 1344 | XP_006718944.1 | |
| XM_011542895.2 | 11 | 1760 | XP_011541197.1 | |
| XM_011542896.2 | 11 | 1569 | XP_011541198.1 | |
| XM_017018000.1 | 11 | 1635 | XP_016873489.1 | |
| XM_017018001.1 | 11 | 1782 | XP_016873490.1 | |
| XM_017018002.1 | 10 | 1291 | XP_016873491.1 | |
| XM_017018003.1 | 10 | 1543 | XP_016873492.1 | |
| XM_017018004.1 | 10 | 1665 | XP_016873493.1 | |
| XM_017018005.1 | 9 | 2898 | XP_016873494.1 | |
| XM_017018006.1 | 10 | 1540 | XP_016873495.1 | |
| AF103801.1 | 11 | 1561 | AAF02421.1 | |
| AF447877.1 | 9 | 1815 | AAQ04652.1 | |
| AK023987.1 | 11 | 1532 | ||
| AK126067.1 | 5 | 2641 | ||
| AK295267.1 | 11 | 1618 | ||
| AK295428.1 | 11 | 1548 | ||
| AK298807.1 | 11 | 1548 | ||
| AK302039.1 | 10 | 1311 | ||
| AL136923.1 | 11 | 1543 | CAB66857.1 | |
| BC002910.2 | 9 | 1140 | AAH02910.2 | |
| BC013902.2 | 11 | 1519 | AAH13902.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556530 | Chr.11:126264724 - 126393990 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map