Assay Details
Target Gene Details
Entrez Gene ID: | 51222 |
Gene Name: | zinc finger protein 219 |
Gene Aliases: |
ZFP219 |
Location: |
Chr.14:21090046-21104704 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF219 | NM_001101672.1 | 4 | 1815 | NP_001095142.1 |
| NM_001102454.1 | 4 | 1805 | NP_001095924.1 | |
| NM_016423.2 | 4 | 1949 | NP_057507.2 | |
| XM_005267739.1 | 4 | 1760 | XP_005267796.1 | |
| XM_006720163.2 | 4 | 1696 | XP_006720226.1 | |
| XM_006720164.3 | 4 | 1698 | XP_006720227.1 | |
| XM_011536825.1 | 4 | 1770 | XP_011535127.1 | |
| XM_017021354.1 | 3 | 1765 | XP_016876843.1 | |
| XM_017021355.1 | 4 | 1661 | XP_016876844.1 | |
| AB015427.1 | 4 | 1939 | BAA90526.1 | |
| AK056261.1 | 2 | 1156 | ||
| AK225811.1 | 4 | 1712 | ||
| AK290700.1 | 4 | 1815 | ||
| BC000694.2 | 3 | 1560 | AAH00694.1 | |
| BC036105.1 | 4 | 1641 | AAH36105.1 | |
| BT006956.1 | 2 | 1522 | AAP35602.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv123n21 | Chr.14:21072704 - 21106531 on Build GRCh38 | Loss |
|
| nsv826882 | Chr.14:21042862 - 21136042 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map