Assay Details
Target Gene Details
Entrez Gene ID: | 83939 |
Gene Name: | eukaryotic translation initiation factor 2A |
Gene Aliases: |
CDA02, EIF-2A, MST089, MSTP004, MSTP089 |
Location: |
Chr.3:150546678-150586016 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF2A | NM_001319043.1 | NP_001305972.1 | ||
| NM_001319044.1 | NP_001305973.1 | |||
| NM_001319045.1 | NP_001305974.1 | |||
| NM_001319046.1 | NP_001305975.1 | |||
| NM_032025.4 | NP_114414.2 | |||
| XM_011513224.1 | XP_011511526.1 | |||
| AF109358.1 | AAQ13506.1 | |||
| AF172818.1 | AAQ13612.1 | |||
| AF212241.1 | AAK14926.1 | |||
| AF497978.1 | AAM83402.1 | |||
| AK027356.1 | BAB55058.1 | |||
| AK293993.1 | ||||
| AK298586.1 | ||||
| AL833101.1 | ||||
| BC011885.1 | AAH11885.1 | |||
| BX647951.1 | ||||
| DA382303.1 |
Target Gene Details
Entrez Gene ID: | 27230 |
Gene Name: | stress-associated endoplasmic reticulum protein 1 |
Gene Aliases: |
RAMP4 |
Location: |
Chr.3:150541993-150603177 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERP1 | XM_011512698.2 | XP_011511000.1 | ||
| AK125413.1 |
More Information
Set Membership: |
|

Genomic Map