Assay Details
Target Gene Details
Entrez Gene ID: | 10106 |
Gene Name: | CTD small phosphatase 2 |
Gene Aliases: |
OS4, PSR2, SCP2 |
Location: |
Chr.12:57819927-57846964 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTDSP2 | NM_005730.3 | NP_005721.3 | ||
| XM_005268556.1 | XP_005268613.1 | |||
| AF000152.1 | AAB71816.1 | |||
| AF022231.1 | AAD09331.1 | |||
| AK226076.1 | ||||
| AK291289.1 | ||||
| AK310183.1 | ||||
| AK310873.1 | ||||
| AK311070.1 | ||||
| AK312413.1 | ||||
| AY279531.1 | AAP34399.1 | |||
| BC065920.1 | AAH65920.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559046 | Chr.12:57768302 - 58084815 on Build GRCh38 | Gain |
|
| nsv726 | Chr.12:57817396 - 57852112 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map