Assay Details
Target Gene Details
Entrez Gene ID: | 64577 |
Gene Name: | aldehyde dehydrogenase 8 family member A1 |
Gene Aliases: |
ALDH12, DJ352A20.2 |
Location: |
Chr.6:134917390-134950122 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH8A1 | NM_001193480.1 | 6 | 1031 | NP_001180409.1 |
| NM_022568.3 | 7 | 1181 | NP_072090.1 | |
| NM_170771.2 | 6 | 1019 | NP_739577.1 | |
| AF303134.1 | 7 | 1172 | AAG42417.1 | |
| AK074266.1 | 6 | 1012 | ||
| AK222848.1 | 6 | 1012 | BAD96568.1 | |
| AK290784.1 | 6 | 966 | ||
| AK298325.1 | 6 | 993 | ||
| BC113862.1 | 7 | 1151 | AAI13863.1 | |
| BC114473.1 | 7 | 1027 | AAI14474.1 | |
| BX647324.1 | 1 | 762 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv604722 | Chr.6:134783086 - 134954927 on Build GRCh38 | Gain |
|
| nsv604723 | Chr.6:134914396 - 135161428 on Build GRCh38 | Gain |
|
| nsv5489 | Chr.6:134903297 - 134948744 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map