Assay Details
Target Gene Details
Entrez Gene ID: | 51069 |
Gene Name: | mitochondrial ribosomal protein L2 |
Gene Aliases: |
CGI-22, MRP-L14, RPML14 |
Location: |
Chr.6:43054029-43059863 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL2 | NM_001300848.1 | NP_001287777.1 | ||
| NM_015950.4 | NP_057034.2 | |||
| XM_005249161.4 | XP_005249218.1 | |||
| AF132956.1 | AAD27731.1 | |||
| AK301041.1 | ||||
| AK314948.1 | ||||
| BC013685.1 | AAH13685.1 | |||
| BC020212.1 | AAH20212.1 | |||
| BM970997.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv462927 | Chr.6:42948182 - 43110718 on Build GRCh38 | Loss |
|
| nsv1017398 | Chr.6:42659705 - 43400998 on Build GRCh38 | Gain |
|
| nsv462930 | Chr.6:43002769 - 43110718 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map